missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ COMMD3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30491
This item is not returnable.
View return policy
Description
COMMD3 Polyclonal antibody specifically detects COMMD3 in Human,Mouse samples. It is validated for Western Blot
Specifications
| COMMD3 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| BUPProtein Bup, C10orf8, chromosome 10 open reading frame 8, COMM domain containing 3, COMM domain-containing protein 3, DKFZp686K0399, FLJ45471, Protein PIL | |
| Between 145 and 195 | |
| 100 μg | |
| Zinc Finger | |
| 23412 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction