missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMMD1, Mouse anti-Human, Clone: 2A12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00150684-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MAAGELEGGKPLSGLLNALAQDTFHGYPGITEELLRSQLYPEVPPEEFRPFLAKMRGILKSIASADMDFNQLEAFLTAQTKKQGGITSDQAAVISKFWKSHKTKIRESLMNQSRWNSGLRGLSWRVDGKSQSRHSAQIHTPVAIIELELGKYGQESEFLCLEFDEVKVNQILKTLSEVEESISTLISQPNSpecifications
| COMMD1 | |
| Monoclonal | |
| Unconjugated | |
| copper metabolism (Murr1) domain containing 1 | |
| C2orf5/MGC27155/MURR1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunoprecipitation, Western Blot | |
| 2A12 | |
| In 1x PBS, pH 7.4 | |
| BC022046 | |
| COMMD1 | |
| COMMD1 (AAH22046.1, 1 a.a. ∼ 190 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 150684 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction