missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMMD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£305.00 - £435.00
Specifications
| Antigen | COMMD1 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18432821
|
Novus Biologicals
NBP1-86797-25ul |
25 μL |
£305.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18495531
|
Novus Biologicals
NBP1-86797 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COMMD1 Polyclonal antibody specifically detects COMMD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| COMMD1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol | |
| 150684 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C2orf5, COMM domain-containing protein 1, copper metabolism (Murr1) domain containing 1, copper metabolism gene MURR1, MGC27155, MURR1chromosome 2 open reading frame 5 (MURR1), Protein Murr1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: AQDTFHGYPGITEELLRSQLYPEVPPEEFRPFLAKMRGILKSIASADMDFNQLEAFLTAQTKKQGGITSDQAAV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title