missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Comfort Rack for Upright Freezers, 130 mm Box Height

Comfort Rack designed to ensure a more ergonomical operating process when working with samples.

Brand:  Tenak™ TE25200

Additional Details : Weight : 2.20000kg

 View more versions of this product

Product Code. 15230864

  • £144.00 / Each
Estimated Shipment: 18-09-2024
to see stock.
Add to basket

Description

Description

  • In the comfort rack, the cryoboxes are placed in drawers
  • Increases the flexibility and reduces the operating weight compared to the rack types
  • Measure the inside height and depth of you freezer when choosing the right rack
  • Racks can be used with all freezer models
  • Constructed of stainless steel
Specifications

Specifications

130 mm (Box)
Upright Freezers
2 x 3
424 x 140 x 268 mm
Boxes (6)
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers