missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Comfort Rack for Chest Freezers, 100 mm Box Height

Product Code. 15220854 Shop All Tenak Products
Change view
Click to view available options
Holds:
Boxes
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
15220854 Boxes Each
15200854 Boxes Each
15280844 Boxes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15220854 Supplier Tenak™ Supplier No. TE24564

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Comfort Rack suitable for use with all chest freezer models.

  • Racks with spring locks mounded on each shelf to secure the cryoboxes
  • Can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate
  • Measure the inside height of your freezer when choosing the right rack
  • Constructed of stainless steel

Specifications

Holds Boxes
Material Stainless Steel
Positions 5
For Use With (Equipment) Chest Frseezers
Dimensions (L x W x H) 140 x 140 x 530 mm
Includes Spring Locks
Height (Metric) 100 mm (Box)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.