Learn More
Abeba™ Comfort 36874 Shoes
Product Code. 15610758
Click to view available options
Size:
36
37
38
39
40
41
42
43
Unit Size:
1 pair
Product Code. 15610758
Brand: Abeba™ 36874/36
Only null left
Add to basket
Add to basket
Stylish hand-sewn seams and PU sole with high slip resistance.
  • CE EN ISO 20347:2012 OB A, E SRB
  • Smooth leather white
  • Slip resistance PU outsole sole
  • Replaceable Reflexor insole with nubs (article no. 3572)
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Instep area with elastic band
  • Buckle fastening
  • Hand-sewn seam
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRB
Color White
Liner Material Microfiber
Material Leather
Size 36
Sole Features Slip-resistant
Sole Material Polyurethane
Style 36874