Learn More
Abeba™ Comfort 36819 Shoes
Product Code. 15690738
Click to view available options
Size:
36
37
38
39
40
41
42
43
Unit Size:
1 pair
Product Code. 15690738
Brand: Abeba™ 36819/40
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Stylish hand-sewn seams and PU sole with high slip resistance.
  • CE EN ISO 20347:2012 OB A, E SRB
  • Smooth leather
  • Black
  • Slip resistance PU outsole sole
  • Replaceable Reflexor insole with nubs (article no. 3572)
  • ESD dissipative according to DIN EN 61340
  • Lining microfiber, respiration absorbing
  • 2 adjustable instep straps
  • Double velcro fastening
  • Hand-sewn seam
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E SRB
Color Black
Liner Material Microfiber
Material Leather
Size 40
Sole Features Slip-resistant
Sole Material Polyurethane
Style 36819