missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for HU13 Midi Horizontal Gel Electrophoresis Unit, 2mm thick
For use with HU13 Midi Horizontal Gel Electrophoresis Unit
Marca: Fisherbrand™
Code : 46
Detalles adicionales : Weight : 2.00000kg
Descripción
- Color coded
- Height adjustable
- Offer complete control over loading volume
- Well depth to maximum 112-sample throughput
Especificaciones
HU13 Midi Horizontal Gel Electrophoresis Unit | |
28 | |
25 μL |
Comb | |
2 mm | |
3 mm |