missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for HE 33 Mini Submarine Unit

Accessory parts for HE 33 Mini Submarine

Brand:  Hoefer™ HE31A-P-1.5

Code : 04

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11569533

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Variety of eight sizes ranging from 1/2 to 16 wells
  • 1 to 1.5 mm Thickness
  • 44/6 to 2.6 mm Well Width
  • 235/35 to 20.5 μL Sample Volume
Specifications

Specifications

HE33 Mini Submarine Unit
Preparative
0.15 cm
1.73/0.2 in.
0.19 in.
Backless Preparative Comb
0.05 in.
250/50 μL
4.4/0.6 cm
0.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers