missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Combination pH Electrodes
Click to view available options
Unit Size:
Each
Specifications
Specifications
| Connector Types | BNC, Pin |
| pH Range | 0 to 12 |
| Type | Combination pH Electrode |
| For Use With (Application) | Fats and creams, soil samples, potable water, semi solid products, low conductivity solutions, emulsions |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction