missing translation for 'onlineSavingsMsg'
Learn More

Draeger Safety Combination Filter 1140 A2B2E2K2HgNO CO 20 P3 R D

Product Code. 15774682
Change view
Click to view available options
:
For Use With: Full Face Mask
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15774682 For Use With: Full Face Mask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15774682 Supplier Draeger Safety Supplier No. 6738814

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Draeger Safety Combination Filter 1140 A2B2E2K2HgNO CO 20 P3 R D can protect against carbon monoxide, mercury vapor, and nitrous gases.

  • Able to cover a wide selection of dangerous gases, contaminants, and particles
  • Filter can only be used in conjunction with full face masks as the weight is more than 300 g
  • Offer clean breathable air for a low cost
  • aluminum housing ensures filters are easily recognisable and are safe from potential damage
  • Shelf life on the combination filters is 6 years (from date of production)
  • Dolomite test passed
  • Approvals include CE approved (EN 141:2000 re. EN 14387, EN 143:2000, EN 371) and SAI Global-approved (Australia)

Specifications

Certifications/Compliance EN 141:2000 re. EN 14387, EN 143:2000, EN 371
Product Type Combination Filter
For Use With (Equipment) Full face mask
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.