missing translation for 'onlineSavingsMsg'
Learn More

Edmund Bühler GmbH™ Combifix KS Rack System

Product Code. 10379141
Click to view available options
Length (Metric):
400 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10379141

Brand: Edmund Bühler GmbH™ 0052 071

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes 3 clamping strips H and rubber mat.

  • Rack system with 3 clamping strips h and rubber mat
  • Shaking platform 400 x 275mm
  • For fastening upright vessels

Order Info

Size

Specifications

Product Type Rack System
Material Rubber
Length (Metric) 400 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.