Learn More
Lovibond™ Combi pack CHLORINE HR / ACIDIFYING GP
Tablet reagent test set
Brand: Lovibond™ 517721BT
Code : NEW
Additional Details : Weight : 0.07020kg
Description
Tablet reagent test set
Combi pack CHLORINE HR / ACIDIFYING GP 100 tests measuring range 0.2 - 16 % NaOCl Potassium iodide method for comparators and photometers.
Specifications
100 Tests | |
CHLORINE 0.2 to 16 % | |
Combi Pack Chlorine HR/Acidifying GP |
Safety and Handling
- combi-pack chlorine HR + acidifying GP
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : citric acid, adipicacid