missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for MultiSUB Maxi Horizontal Gel Electrophoresis Unit, 2mm Thick

For use in preparatory electrophoresis and high-throughput screening.

Brand:  Fisherbrand™

Code : 46

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11843453

  • £28.30 / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket

Description

Description

  • Recommended to scale-up nucleic acid volumes for preparatory techniques
  • Maximise sample volume
  • Height adjustable to give user full control over well depth and sample-loading volume
  • Rigid back which prevents heat-induced warping
  • Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
  • 20-sample and 40-sample combs are Multichannel pipettor compatible
Specifications

Specifications

MultiSUB Maxi Horizontal Gel Electrophoresis Unit
36
2 mm
Comb
1 Each
29 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers