missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for TV100 Standard Twin-Plate Mini-Gel Vertical Electrophoresis Unit
For use with TV100 Standard Twin-Plate Mini-Gel Vertical Electrophoresis Unit
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 5.00000kg
Description
- Available in 0.75, 1, 1.5 and 2mm thickness
Specifications
TV100 Standard Twin-Plate Mini-Gel Vertical Electrophoresis Unit | |
Teeth: 15 mm | |
2 mm | |
3.75 mm |
Comb | |
12 | |
112 μL |