missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FosB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568534
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 91%.
c-FOS is a nuclear phosphoprotein which forms a tight but non-covalently linked complex with the JUN/AP-1 transcription factor. It has a critical function in regulating the development of cells destined to form and maintain the skeleton, and is thought to have an important role in signal transduction, cell proliferation and differentiation.
Specifications
| FosB | |
| Polyclonal | |
| Unconjugated | |
| Fosb | |
| activator protein 1; AP-1; DKFZp686C0818; FBJ murine osteosarcoma viral oncogene homolog B; FBJ murine osteosarcoma viral oncogene-like protein B; FBJ osteosarcoma oncogene B; FBJ osteosarcoma viral oncogene isoform deltaFosb-2; fos b; FOSB; FosB proto-oncogene, AP-1 trancription factor subunit; FosB proto-oncogene, AP-1 transcription factor subunit; fra-2; G0/G1 switch regulatory protein 3; G0S3; GOS3; GOSB; LOW QUALITY PROTEIN: protein fosB; MGC42291; oncogene FOS-B; protein fosB; zgc:92354 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 100360880, 2354 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| D3ZLB7, P53539 | |
| Fosb | |
| synthetic peptide directed towards the N-terminal region of rat FosB. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction