missing translation for 'onlineSavingsMsg'
Learn More

Coluracetam, MedChemExpress

Product Code. 16439867
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16439867 5 mg 5mg
16437366 100 mg 100mg
16427398 10 mg 10mg
16413427 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16439867 Supplier Medchem Express Supplier No. HY17553

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Coluracetam(MKC-231) is a new choline uptake enhancer.

Coluracetam(MKC-231) is a new choline uptake enhancer.

Specifications

Chemical Name or Material Coluracetam
CAS 135463-81-9
Color White
Purity Grade Notes Research
Molecular Formula C19H23N3O3
Quantity 5 mg
Synonym MKC-231
Solubility Information DMSO : ≥ 20 mg/mL (58.58 mM) ∣H2O : 0.4 mg/mL (1.17 mM; Need ultrasonic and warming)
SMILES O=C(NC1=C2C(OC(C)=C2C)=NC3=C1CCCC3)CN4C(CCC4)=O
Molecular Weight (g/mol) 341.4
Formula Weight 341.4
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.