missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Columbia Blood Agar Base
Description
- Columbia Blood Agar is an infusion-free basal medium to use with or without blood for the cultivation of fastidious microorganisms.
- Used with blood in isolating and cultivating fastidious microorganisms
- Used in qualitative procedures
- For isolating and cultivating Haemophilus species from clinical specimens
- Columbia Blood Agar is utilized as the base for media containing blood and for selective media formulations in which various combinations of antimicrobial agents are used as additives.
- Columbia Blood Agar Base EH (Enhanced Hydrolysis) is used with blood in isolating and cultivating fastidious microorganisms.
Specifications
Specifications
| Certifications/Compliance | AOAC, BAM, EP, ISO |
| Form | Dehydrated |
| Media Type | Columbia Blood Agar Base |
| For Use With (Application) | For Enhanced Beta-Hemolytic Reactions after Overnight Incubation and for Cultivation of Fastidious Microorganisms |
| Packaging Type | Pail |
| Quantity | 10 kg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?