missing translation for 'onlineSavingsMsg'
Learn More

Columbamine, MedChemExpress

Product Code. 16409208
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16409208 10 mg 10mg
16401618 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16409208 Supplier Medchem Express Supplier No. HYN0926
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Columbamine is a quaternary isoquinoline alkaloid isolated from Argemone mexicana.

Columbamine is a quaternary isoquinoline alkaloid isolated from Argemone mexicana.

Specifications

Chemical Name or Material Columbamine
CAS 3621-36-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H20NO4
Quantity 10 mg
Synonym Columbamin Dehydroisocorypalmine
Solubility Information DMSO : ≥ 23 mg/mL (67.97 mM)
SMILES COC1=C(OC)C2=C[N+]3=C(C4=CC(O)=C(OC)C=C4CC3)C=C2C=C1
Molecular Weight (g/mol) 338.38
Formula Weight 338.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.