missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ratiolab™ Colorless -E 100 - 1200 μL Pipette Tips
Description
- Uniform high quality and precision makes these tips fully effective
- Can be used with specially developed Multirack, Refill Systems, and Multiboxes
- Provides for simple and safe pipetting
- Can be used in the most sensitive applications
- Provided in packs of 1000
Specifications
Specifications
| Volume (Metric) | 100 to 1200 μL |
| Color | Clear |
| Format | Bulk |
| Quantity | x 1000 |
| Product Type | Pipette tips |
| For Use With (Equipment) | Universal Fit, Both Manual or Electronic, Universal Fit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?