missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Colorimeter Accessories for AF710-3 Tintometer
£13.65
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11857474
|
Lovibond™
122340 |
Tungsten Halogen Lamp |
£13.65
Each |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
For use with Lovibond Colorimeter (09-053-362).
Accessories:
- Sample Tubes: pair of glass sample tubes to AOCS requirements
- Tunsten Halogen Lamp: 12V 20w
- Drip Tray: For use with porcelain (09-053-366)
- Porcelain White Reference: for AOCS; measures 85 x 170mm
Specifications
Lovibond AF710-3 Tintometer |