missing translation for 'onlineSavingsMsg'
Learn More

Primary Color Solution, Yellow, EP Grade, Reagecon™
BIOPHARMA

Product Code. 11845853 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11845853 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11845853 Supplier Reagecon Supplier No. EPYS01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to calibrate, control, qualify and validate color measurement instruments. Primary Color Solution, Yellow, EP Grade, Reagecon™ is a ready-to-use colorimetric solution presented in high-quality, tamper-evident bottles. It is prepared with iron chloride in accordance with Eu. Ph. Chapter 2.

This Primary Color Solution is a ready-to-use coloration standard presented in high-quality, tamper-evident bottles for use as a calibration and/or quality control standard. It is prepared with iron (III) chloride, hydrochloric acid and water.
  • Manufactured according to European Pharmocopoeia Chapter 2 guidelines with a Balance Certificate of Calibration issued in accordance with ISO 17025
  • Prepared gravimetrically on a weight/weight basis using OIML traceable weights
  • Concentration verified using a high-performance spectrophotometer calibrated using ISO Guide 34 accredited spectrophotometric standards
  • The Primary Color Solutions can be combined in precise ratios to form the Standard Color Solutions

Specifications

Description European Pharmacopoeia Primary Color Solution
CAS 7705-08-0
Analyte or Component Names Iron (III) chloride, Hydrochloric acid and water
Physical Form Liquid
Molecular Formula Cl3Fe
MDL Number MFCD00011005
Color Yellow
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]
Element Iron (III) Chloride, Hydrochloric Acid
IUPAC Name iron(3+) trichloride
Molecular Weight (g/mol) 162.20
Quantity 100 mL
Formula Weight 162.20g/mol
Chemical Name or Material Iron(III) Chloride
Show More Show Less
compliance-icons
Product Identifier
  • PRIMARY YELLOW SOLUTION
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: iron trichloride, hydrochloric acid 37%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.