missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 4.00, Red, Reagecon™
BIOPHARMA

Product Code. 11421974 Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Certified by an ISO 17025 accredited method and NIST traceable. Reagecon™ Buffer Solution, pH 4.00, Red is tested at 20°C to an accuracy of ±0.01. This calibration and control buffer is specially formulated to ensure its stability, is color coded and features a twin-neck bottle design.

Color coded and packaged for convenience

  • Colored red for easy identification of pH value
  • Supplied in a Twin Neck bottle that features an integrated calibrating chamber

To ensure reliable, reportable pH measurements, this pH buffer standard is directly traceable to the IUPAC pH scale by an unbroken chain of traceability. Traceable analysis is necessary for consistency and universal acceptance of pH results, including acceptance by regulatory bodies.

  • Traceable to SRM for NIST
  • Certification to ISO 9001:2008
  • Accredited to ISO 17025

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Red
pH 4.00 ±0.01
Physical Form Liquid
Quantity 500 mL
Solution Type Colored pH Buffer
Odor Characteristic
Packaging Twin Neck bottle with integrated calibrating chamber
Traceability to NIST Yes
Solubility Information Fully miscible in water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.