missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Colon Matched Tumor and Normal Tissue Slides (Tumor)- Paraffin

Product Code. 18627161 Shop All Bio Techne Products
Click to view available options
Quantity:
10 Slides
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 18627161

Brand: Novus Biologicals™ NBP277978

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tissue Slides

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 Slides
For Use With (Application) Immunohistochemistry (IHC), Immunohistochemistry (Paraffin), ISH
Species Human
Target Specificity Colon
Content And Storage Store at 4°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.