missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Collibri™ Library Quantification Kit
Shop All Thermo Scientific Products
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
500 reactions
Each
Description
- Includes Platinum II Taq Hot Start DNA Polymerase and enables convenient and accurate quantification of NGS libraries
- Contains all components needed for the amplification reaction, including primers and passive reference dye, and does not require any preparation before use
- Due to the universal concentration of the passive reference dye in the master mix, the kit can be used with any qPCR instrument without adjustments
- Accurate quantification of libraries made from various sample types, including degraded samples
- Ease of use with all components ready to use -- no reagent preparation required
- Visual cues - specially formulated dyes included to track pipetting steps
- Platinum hot-start technology - superior specificity, sensitivity, and room-temperature reaction setup Unmatched convenience
- Contains:
- Platinum II Taq Hot Start DNA Polymerase
- Optimized buffer
- dNTPs
- Primer mix targeting adapter sequences
- SYBR green dye
- Passive reference dye
- Blue dye to track pipetting steps
- Unique formulation provides ease of use at every step of the workflow:
- No reagent preparation
- Tracking of pipetting steps
- One cycling protocol for libraries of various lengths and GC/AT-content levels
- Universal passive reference dye concentration - a single master mix for all qPCR instruments
- Also includes a ready-to-use library dilution buffer that contains stabilizing agents to enable library integrity after dilution. The buffer contains yellow dye to track pipetting steps. When combined, the blue master mix and yellow diluted libraries become a green solution. This feature eases tracking of the addition of the diluted libraries or DNA standards to the master mix in the qPCR plate wells, helping reduce pipetting errors.
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Detection Method | SYBR |
| Format | Kit |
| Sample Type | DNA, RNA |
| For Use With (Equipment) | 7500 Fast System, 7900HT Fast System, QuantStudio™, ViiA™ 7 System, StepOne™ System, StepOnePlus™ System |
| GC-Rich PCR Performance | Low |
| For Use With (Application) | Illumina NGS Library Quantification |
| No. of Reactions | 100 Reactions |
| PCR Method | qPCR |
| Polymerase | Platinum™ II Taq Hot Start DNA Polymerase |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction