missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Collectrin/TMEM27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30745
This item is not returnable.
View return policy
Description
Collectrin/TMEM27 Polyclonal antibody specifically detects Collectrin/TMEM27 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Collectrin/TMEM27 | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| NP_065716.1 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| collectrin, kidney-specific membrane protein, NX-17, transmembrane protein 27NX170610008J07Rik | |
| Produced in rabbits immunized with purified, recombinant Human Collectrin/TMEM27 (RefSeq#: NP_065716.1; Met1-Pro141) | |
| 100 μL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 57393 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction