Learn More
STEMCELL Technologies Collagenase/Hyaluronidase
Product Code. 17139911
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
Product Code. Quantity unitSize
17139911 10 mL 10mL
1 options
Product Code. 17139911 Supplier STEMCELL Technologies Supplier No. 07912
10X Collagenase/Hyaluronidase in Dulbecco's Modified Eagle's Medium (DMEM)
  • 10X Collagenase/Hyaluronidase in Dulbecco's Modified Eagle's Medium (DMEM) for the enzymatic dissociation of human mammary tissue
Specifications
Cell Type Mammary Cells, Other, Prostate Cells
Quantity 10 mL