missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen XXV alpha 1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-248PE
This item is not returnable.
View return policy
Description
Collagen XXV alpha 1 Polyclonal antibody specifically detects Collagen XXV alpha 1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Collagen XXV alpha 1 | |
| Polyclonal | |
| Western Blot | |
| Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component | |
| A synthetic peptide mapping to residues 155-169 [KGEQGDQGPRMVFPK] of the NC2-2 region of human and mouse CLAC-P. | |
| 0.1 mL | |
| Neuroscience | |
| 84570 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur