missing translation for 'onlineSavingsMsg'
Learn More

Collagen XXV alpha 1 Antibody (1D2), Novus Biologicals™

Product Code. 18401999 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401999 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401999 Supplier Novus Biologicals Supplier No. H00084570M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Collagen XXV alpha 1 Monoclonal antibody specifically detects Collagen XXV alpha 1 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen XXV alpha 1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1D2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_942014
Gene Alias Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component
Host Species Mouse
Immunogen COL25A1 (NP_942014.1, 57 a.a. ∽ 122 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NDLQARIAALESAKGAPSIHLLPDTLDHLKTMVQEKVERLLAQKSYEHMAKIRIAREAPSECNCPA
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84570
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.