missing translation for 'onlineSavingsMsg'
Learn More

Collagen XVII Rabbit anti-Human, Clone: 8W2C8, Novus Biologicals™

Product Code. 18300045 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18300045 20 μg 20µL
18346766 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18300045 Supplier Novus Biologicals Supplier No. NBP31648520UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Collagen XVII Monoclonal antibody specifically detects Collagen XVII in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen XVII
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8W2C8
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 180 kDa bullous pemphigoid antigen 2, BA16H23.2, BP180alpha 1 type XVII collagen, BPAG2bA16H23.2 (collagen, type XVII, alpha 1 (BP180)), Bullous pemphigoid antigen 2, bullous pemphigoid antigen 2 (180kD), Collagen 17, collagen alpha-1(XVII) chain, collagen XVII, alpha-1 polypeptide, collagen, type XVII, alpha 1, Collagen-17, FLJ60881, KIAA0204, LAD-1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1300-1400 of human Collagen XVII (Q9UMD9).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1308
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.