missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen XVII Antibody (15G9), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Collagen XVII |
|---|---|
| Clone | 15G9 |
| Dilution | Western Blot 1:500-1:2000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043773
|
Novus Biologicals
NBP3-26647-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043134
|
Novus Biologicals
NBP3-26647-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Collagen XVII Monoclonal antibody specifically detects Collagen XVII in Human samples. It is validated for Western Blot, ELISASpecifications
| Collagen XVII | |
| Western Blot 1:500-1:2000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 180 kDa bullous pemphigoid antigen 2, BA16H23.2, BP180alpha 1 type XVII collagen, BPAG2bA16H23.2 (collagen, type XVII, alpha 1 (BP180)), Bullous pemphigoid antigen 2, bullous pemphigoid antigen 2 (180kD), Collagen 17, collagen alpha-1(XVII) chain, collagen XVII, alpha-1 polypeptide, collagen, type XVII, alpha 1, Collagen-17, FLJ60881, KIAA0204, LAD-1 | |
| A synthesized peptide derived from Human Collagen XVII [UniProt Q9UMD9] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 15G9 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 1308 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title