missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen XI alpha 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | Collagen XI alpha 2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Collagen XI alpha 2 Polyclonal specifically detects Collagen XI alpha 2 in Human samples. It is validated for Western Blot.Specifications
| Collagen XI alpha 2 | |
| Polyclonal | |
| Rabbit | |
| P13942 | |
| 1302 | |
| Synthetic peptides corresponding to COL11A2(collagen, type XI, alpha 2) The peptide sequence was selected from the N terminal of COL11A2. Peptide sequence TADRFQAEEYGEGGTDPPEGPYDYTYGYGDDYREETELGPALSAETAHSG. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| collagen alpha-2(XI) chain, collagen, type XI, alpha 2, DFNA13, DFNB53, HKE5, PARP, pro-a2 chain of collagen type XI, STL3 | |
| COL11A2 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title