missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen X alpha 1 Rabbit anti-Mouse, Rat, Clone: 0F5Q9, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | Collagen X alpha 1 |
|---|---|
| Clone | 0F5Q9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18356502
|
Novus Biologicals
NBP3-15419-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18397756
|
Novus Biologicals
NBP3-15419-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Collagen X alpha 1 Monoclonal antibody specifically detects Collagen X alpha 1 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| Collagen X alpha 1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1300 | |
| IgG | |
| Affinity purified |
| 0F5Q9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| COL10A1, Collagen Alpha-1(X) Chain, Collagen X, Alpha-1 Polypeptide, Schmid Metaphyseal Chondrodysplasia | |
| A synthetic peptide corresponding to a sequence within amino acids 500-600 of human Collagen X alpha 1 (Q03692). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title