missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VI alpha 3 Antibody (118), Alexa Fluor™ 647, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Collagen VI alpha 3 Monoclonal specifically detects Collagen VI alpha 3 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Collagen VI alpha 3 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 118 |
| Conjugate | Alexa Fluor 647 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Collagen Alpha-3(VI) Chain, Collagen VI, Alpha-3 Polypeptide, collagen, type VI, alpha 3 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Collagen VI alpha 3 (P12111-1; 3101T-3177T). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?