missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VI alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38600-100ul
This item is not returnable.
View return policy
Description
Collagen VI alpha 1 Polyclonal antibody specifically detects Collagen VI alpha 1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Collagen VI alpha 1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| Collagen 6, collagen alpha-1(VI) chain, collagen VI, alpha-1 polypeptide, collagen, type VI, alpha 1, Collagen-6, human mRNA for collagen VI alpha-1 C-terminal globular domain10alpha 1 (VI) chain (61 AA), OPLL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 19-250 of human Collagen VI alpha 1 (NP_001839.2).,, Sequence:, AQDEPETPRAVAFQDCPVDLFFVLDTSESVALRLKPYGALVDKVKSFTKRFIDNLRDRYYRCDRNLVWNAGALHYSDEVEIIQGLTRMPGGRDALKSSVDAVKYFGKGTYTDCAIKKGLEQLLVGGSHLKENKYLIVVTDGHPLEGYKEPCGGLEDAVNEAKHLGVKVFSVAITPDHLEPRLSIIATDHTYRRNFTAADWGQSRDAEEAISQTIDTIVDMIKNNVEQVCCSF | |
| 100 μL | |
| Cell Biology, Cellular Markers, Extracellular Matrix | |
| 1291 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction