missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Collagen V Native Protein
Shop All Bio Techne Products
Click to view available options
:
0.5mL; Unlabeled
Description
Native COL5A1. Source: Human Placenta (Accession #: P20908) The Collagen V Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.
Specifications
Specifications
| Accession Number | P05997 |
| For Use With (Application) | ELISA, Immunohistochemistry, SDS-PAGE, Western Blot |
| Formulation | 0.5M Acetic Acid |
| Gene ID (Entrez) | 1289 |
| Name | Collagen V protein |
| Source | Human serum |
| Storage Requirements | 4°C. Do not freeze. |
| Cross Reactivity | Human |
| Purity or Quality Grade | Chromatography |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction