missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen V Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-26552
This item is not returnable.
View return policy
Description
Collagen V Polyclonal specifically detects Collagen V in Human, Bovine, Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Dot Blot.
Specifications
| Collagen V | |
| Polyclonal | |
| Biotin | |
| PBS with Sodium Azide | |
| COL5A1 | |
| Goats were hyperimmunized with human type V collagen. | |
| 0.1 mg | |
| Primary | |
| Reacts with conformational determinants on human type V collagen as demonstrated by ELISA. May react with type V collagen from other species. Exhibits <10% cross reactivity with collagen type I, II, III, IV and VI. The antibody has not been tested for reactivity with other ECM proteins (e.g., laminin, fibronectin). | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 0.4mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:1000-1:4000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:20, Immunohistochemistry-Frozen 1:20-1:40, Dot Blot 1:100-1:2000, Immunocytochemistry | |
| alpha 1 type V collagen, Collagen 5, collagen alpha-1(V) chain, collagen, type V, alpha 1, Collagen-5, EC 2.7.7.6, EC 6.1.1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1289 | |
| Human, Bovine, Equine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction