missing translation for 'onlineSavingsMsg'
Learn More

Collagen V Antibody (1E2-E4/Col5), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18425115 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425115 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425115 Supplier Novus Biologicals Supplier No. NBP105118MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Collagen V Monoclonal antibody specifically detects Collagen V in Human, Porcine, Bovine, Canine, Kangaroo, Rabbit, Sheep, Chicken (Negative), Guinea Pig (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen V
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1E2-E4/Col5
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate
Gene Alias alpha 1 type V collagen, Collagen 5, collagen alpha-1(V) chain, collagen, type V, alpha 1, Collagen-5, EC 2.7.7.6, EC 6.1.1
Host Species Mouse
Immunogen Acid-digested pepsin soluble dog type V collagen
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1289
Target Species Human, Pig, Bovine, Canine, Kangaroo
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.