missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen V Antibody (1E2-E4/Col5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05118AF647
This item is not returnable.
View return policy
Description
Collagen V Monoclonal specifically detects Collagen V in Human, Porcine, Bovine, Canine, Kangaroo, Rabbit, Sheep, Chicken (Negative), Guinea Pig (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Collagen V | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| COL5A1 | |
| Acid-digested pepsin soluble dog type V collagen | |
| 0.1 mL | |
| Cytoskeleton Markers, Extracellular Matrix | |
| 1289 | |
| Human, Pig, Bovine, Canine, Kangaroo, Rabbit, Sheep, Chicken (Negative), Guinea Pig (Negative), Mouse (Negative) | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1E2-E4/Col5 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| alpha 1 type V collagen, Collagen 5, collagen alpha-1(V) chain, collagen, type V, alpha 1, Collagen-5, EC 2.7.7.6, EC 6.1.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| It has been shown to have no cross reactivity with type I, III and VI collagens by ELISA and immunoblotting. There is no evidence for cross-reactivity with other connective tissue proteins (laminin, fibronectin, elastin). | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction