missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV Antibody (M3F7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08515H
This item is not returnable.
View return policy
Description
Collagen IV Monoclonal antibody specifically detects Collagen IV in Human, Rat, Avian samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Collagen IV | |
| Monoclonal | |
| HRP | |
| BSVD;BSVD1;COL4A1s;Collagen alpha-1(IV) chain;PADMAL;RATOR | |
| alpha1 (IV)-alpha2(IV) triple helix. The epitope is triple helical domain about 900 from the N-terminus | |
| 100 μg | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
| 1282 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| M3F7 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Avian | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction