missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-26550
This item is not returnable.
View return policy
Description
Collagen IV Polyclonal specifically detects Collagen IV in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.
Specifications
| Collagen IV alpha 1 | |
| Polyclonal | |
| Biotin | |
| PBS with Sodium Azide | |
| COL4A1 | |
| Goats were hyperimmunized with human type IV collagen. | |
| 0.1 mg | |
| Stem Cell Markers | |
| 1282 | |
| Human, Rat, Monkey, Monkey | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.4mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:1000-1:4000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:20, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:20-1:40, Dot Blot 1:100-1:2000, Immunocytochemistry | |
| alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reacts with conformational determinants on human type IV collagen as demonstrated by ELISA. May react with type IV collagen from other species. Exhibits <10% cross reactivity with collagen type I, II, III, V and VI. The antibody has not been tested for reactivity with other ECM proteins (e.g., laminin, fibronectin). | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction