missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen III alpha 1/COL3A1 Antibody [Janelia Fluor™ 585], Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-26547JF585
This item is not returnable.
View return policy
Description
Collagen III alpha 1/COL3A1 Polyclonal antibody specifically detects Collagen III alpha 1/COL3A1 in Human samples. It is validated for ELISA
Specifications
| Collagen III alpha 1/COL3A1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Janelia Fluor 585 | |
| alpha1 (III) collagen, alpha-1 type III collagen, COL3A1, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, Collagen-III, collagenIII, EDS4A, EDSVASC, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534, PMGEDSV | |
| This Collagen III alpha 1/COL3A1 Antibody was developed by hyperimmunizing goats with human type III collagen. | |
| 0.1 mL | |
| Cancer, Cell Biology, Extracellular Matrix | |
| 1281 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction