missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen III alpha 1/COL3A1 Antibody, DyLight 594, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-26547DL594
This item is not returnable.
View return policy
Description
Collagen III alpha 1/COL3A1 Polyclonal specifically detects Collagen III alpha 1/COL3A1 in Human samples. It is validated for ELISA.
Specifications
| Collagen III alpha 1/COL3A1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| alpha1 (III) collagen, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, EDS4A, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534 | |
| Goat | |
| 139 kDa | |
| 0.1 mL | |
| Cancer, Cell Biology, Extracellular Matrix | |
| Reacts with conformational determinants on human type III collagen as demonstrated by ELISA. May react with type III collagen from other species. Exhibits <10% cross reactivity with collagen type I, II, IV, V and VI. The antibody has not been tested for reactivity with other ECM proteins (e.g., laminin, fibronectin). | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| COL3A1 | |
| This Collagen III alpha 1/COL3A1 Antibody was developed by hyperimmunizing goats with human type III collagen. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction