missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen III alpha 1/COL3A1 Antibody (1E7-D7/Col3), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
£540.00
Specifications
| Antigen | Collagen III alpha 1/COL3A1 |
|---|---|
| Clone | 1E7-D7/Col3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 610 SE |
Description
Collagen III alpha 1/COL3A1 Monoclonal antibody specifically detects Collagen III alpha 1/COL3A1 in Human, Mouse, Rat, Canine, Kangaroo, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| Collagen III alpha 1/COL3A1 | |
| Western Blot | |
| mFluor Violet 610 SE | |
| Mouse | |
| Cancer, Cell Biology, Extracellular Matrix | |
| 50mM Sodium Borate | |
| 1281 | |
| IgG1 κ | |
| Protein A or G purified |
| 1E7-D7/Col3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| alpha1 (III) collagen, alpha-1 type III collagen, COL3A1, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, Collagen-III, collagenIII, EDS4A, EDSVASC, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534, PMGEDSV | |
| This Collagen III alpha 1/COL3A1 Antibody (1E7-D7/Col3) was developed against acid-digested pepsin soluble human type III collagen | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title