missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen III alpha 1/COL3A1 Antibody (1E7-D7/Col3), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Collagen III alpha 1/COL3A1 Monoclonal antibody specifically detects Collagen III alpha 1/COL3A1 in Human, Mouse, Rat, Canine, Kangaroo, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | Collagen III alpha 1/COL3A1 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 1E7-D7/Col3 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Formulation | 50mM Sodium Borate |
| Gene Alias | alpha1 (III) collagen, alpha-1 type III collagen, COL3A1, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, Collagen-III, collagenIII, EDS4A, EDSVASC, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534, PMGEDSV |
| Host Species | Mouse |
| Immunogen | This Collagen III alpha 1/COL3A1 Antibody (1E7-D7/Col3) was developed against acid-digested pepsin soluble human type III collagen |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?