missing translation for 'onlineSavingsMsg'
Learn More

Collagen II Antibody, Novus Biologicals™

Product Code. 18386941 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18386941 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18386941 Supplier Novus Biologicals Supplier No. NBP1777950.05MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 4 publications

Collagen II Polyclonal specifically detects Collagen II in Human, Mouse, Rat, Bovine, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen II
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, ELISA 1:5000-1:50000, Immunohistochemistry 1:50-1:400, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:10-1:500
Formulation 0.125 M sodium borate, 0.075 M Sodium Chloride, 0.005 M EDTA, pH 8.0 with 0.01% sodium azide
Gene Accession No. P02458
Gene Alias Alpha-1 type II collagen, ANFH, AOM, cartilage collagen, chondrocalcin, COL11A3, Collagen 2, collagen alpha-1(II) chain, collagen II, alpha-1 polypeptide, collagen type 2, collagen, type II, alpha 1, collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphysealdysplasia, congenital), Collagen-2, MGC131516, SEDC
Gene Symbols COL2A1
Host Species Rabbit
Immunogen Collagen Type II from human knee cartilage and bovine nasal cartilage.. Uniprot ID P02458
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 1280
Test Specificity Typically less than 1% cross reactivity against other types of collagens was detected by ELISA against purified standards. Some class specific anti-collagens may be specific for three-dimensional epitopes which may result in diminished reactivity with de
Target Species Human, Mouse, Rat, Avian, Bovine, Canine, Sheep, Silkworm
Content And Storage Store at 4°C short term. For extended storage, add an equal volume of glycerol, aliquot and store at -20°C or below. Avoid repeated freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.