missing translation for 'onlineSavingsMsg'
Learn More

Collagen II Antibody (M2139), Novus Biologicals™

Product Code. 18335785 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335785 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335785 Supplier Novus Biologicals Supplier No. NBP311228

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Collagen II Monoclonal specifically detects Collagen II in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Immunohistochemistry-Paraffin, Functional, Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen II
Applications Immunohistochemistry (Paraffin), Functional Assay, Immunoassay
Classification Monoclonal
Clone M2139
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin, Functional, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Alpha-1 type II collagen, ANFH, AOM, cartilage collagen, chondrocalcin, COL11A3, Collagen 2, collagen alpha-1(II) chain, collagen II, alpha-1 polypeptide, collagen type 2, collagen, type II, alpha 1, collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphysealdysplasia, congenital), Collagen-2, MGC131516, SEDC
Host Species Mouse
Immunogen The monoclonal antibody M2139 reacts with the J1 epitope (triple helical position 551-564) of collagen type II.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1280
Target Species Human, Mouse, Rat, Bovine, Chicken
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.