missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen II Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-46876
This item is not returnable.
View return policy
Description
Collagen II Polyclonal antibody specifically detects Collagen II in Human, Mouse, Rat, Bovine, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Collagen II | |
| Polyclonal | |
| Western Blot 1:5000-1:10000, Flow Cytometry 1:10 - 1:1000, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:100 | |
| P02458 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Reconstitute with 50 μg deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| FITC | |
| Lyophilized from 0.01 M Sodium Phosphate, 0.25 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
| Alpha-1 type II collagen, ANFH, AOM, cartilage collagen, chondrocalcin, COL11A3, Collagen 2, collagen alpha-1(II) chain, collagen II, alpha-1 polypeptide, collagen type 2, collagen, type II, alpha 1, collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphysealdysplasia, congenital), Collagen-2, MGC131516, SEDC | |
| Collagen II from human knee cartilage and bovine nasal cartilage. (Uniprot: P02458) | |
| 0.05 mg | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
| 1280 | |
| Human, Mouse, Rat, Bovine, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction