missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen II Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-21663
This item is not returnable.
View return policy
Description
Collagen II Polyclonal specifically detects Collagen II in Human, Mouse, Rat, Bovine, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Collagen II | |
| Polyclonal | |
| Biotin | |
| Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
| Alpha-1 type II collagen, ANFH, AOM, cartilage collagen, chondrocalcin, COL11A3, Collagen 2, collagen alpha-1(II) chain, collagen II, alpha-1 polypeptide, collagen type 2, collagen, type II, alpha 1, collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphysealdysplasia, congenital), Collagen-2, MGC131516, SEDC | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Some class specific anti-collagens may be specific for three-dimensional epitopes which may result in diminished reactivity with denatured collagen or formalin-fixed, paraffin embedded tissues. This antibody reacts with most mammalian Collagen II and has expected cross-reactivity with Type IV and VI and negligible cross reactivity to Type I, III and V collagens. Non-specific cross reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins has not been tested. | |
| Human, Mouse, Rat, Bovine, Sheep, Canine, Silkworm | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:5000-1:10000, ELISA 1:10000-1:50000, Immunohistochemistry 1:200-1:1000, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| P02458 | |
| COL2A1 | |
| Collagen II from human knee cartilage and bovine nasal cartilage. (Uniprot: P02458) | |
| 0.1 mg | |
| Cytoskeleton Markers, Extracellular Matrix, Stem Cell Markers | |
| 1280 | |
| Reconstitute with 100 ul deionized water (or equivalent) | |
| Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction