missing translation for 'onlineSavingsMsg'
Learn More

Collagen II Antibody (819447) [DyLight 488], Novus Biologicals™

Product Code. 30730930 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30730930 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30730930 Supplier Novus Biologicals Supplier No. FAB36151K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Collagen II Monoclonal antibody specifically detects Collagen II in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen II
Applications ELISA
Classification Monoclonal
Clone 819447
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Alpha-1 type II collagen, ANFH, AOM, cartilage collagen, chondrocalcin, COL11A3, Collagen 2, collagen alpha-1(II) chain, collagen II, alpha-1 polypeptide, collagen type 2, collagen, type II, alpha 1, collagen, type II, alpha 1 (primary osteoarthritis, spondyloepiphysealdysplasia, congenital), Collagen-2, MGC131516, SEDC
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Pro-Collagen II, Gln26-Arg266 and Gly957-Leu1487, Accession # P02458
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1280
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.